log in or register
48-pin Insert for Labconco Glassware Washers
EW-98963-63
$162.00 USD/EACH
Usually ships in 14 days.
Qty
  • Customize your glassware washer with these accessories
This 48-pin insert accommodates a variety of glassware for washing. The 304 stainless steel construction includes plastic tips to protect glassware from scratching. Pins hold glassware in place during wash cycle.
Manufacturer number 4591601
Model 4591601
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title