United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 6
of
10 >
Representative photo only
50 mL self standing tube PPCO Non sterile 20 bags of 25
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-06344-25
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Centrifuges
>
Centrifuge Accessories
>
Centrifuge Tubes
>
Cole-Parmer Conical-Bottom Disposable Plastic Centrifuge Tubes
Share
|
50 mL self-standing tube, PPCO, Non-sterile, 20 bags of 25
EW-06344-25
$201.00 USD/CASE
Quantity Discounts
Order
Discount
Price
[X]
6
10%
$180.90/CASE
30
15%
$170.85/CASE
120
20%
$160.80/CASE
In stock.
Qty
:
Add to Cart
Save to shopping list
Ideal for applications where other tubes would shatter
Cole Parmer tubes are available in polystyrene and polypropylene co-polymer (PPCO). The 50-ml tubes 06344-25 and 06344-26 are self-standing with molded-in graduations; all other models feature blue printed graduations and a writing patch. Tubes are available sterile-packed in trays or bags, or nonsterile-packed with caps separate.
Specifications
Product Type
Conical-bottom Tubes
Sterile
Yes
Cap
Flat top/rim seal
Color
clear
Dimensions (OD (mm))
29
Dimensions (L (mm))
115
Capacity (mL)
50
Graduations
Yes
Autoclavable
Yes
Packaging
20 bags of 25
Qty/cs
500
Material
Polypropylene
Brand
Cole-Parmer
Manufacturer number
3264
Model
3264
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
High Throughput Maxi-Prep Using the New Thermo Scientific General Purpose Centrifuge with Feberlite Rotors
Temperature Compatibility Guidelines for Centrifuge Tubes and Bottles
Isolating Genomic DNA from Whole Blood
Show All...
Common Centrifuge Terms
Basics of Centrifugation
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title