Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

55 Gallon, Full Coverage, Plastic Drum Heater, Dual Zone - 240 V

BriskHeat  |   Item# EW-09300-55

$1,032.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Usually ships in 14 days.
Qty

  • Provides safe, uniform heat to 55-gallon metal or plastic drums
  • Built-in adjustable thermoswitch prevents temperature runaway
  • Lightweight flexible design makes installation quick and easy
These insulated drum heaters provide a safe and efficient method of temperature and viscosity control for liquids in 55-gallon drums. Simply wrap insulated drum heater around a 55-gallon drum, adjust the built-in thermostat to desired temperature setting, and uniform heat is evenly distributed around the entire drum. The grounded heating element provides a high-watt density, ensuring rapid heating times—one inch thick fiberglass insulation prevents heat loss. Prevent further heat loss by ordering the drum insulation top cover (optional) below.
Drum heaters feature a VELCRO® seal for easy installation and removal and models are available for both metal and plastic 55-gallon drums. Heaters come with standard three-prong plug (240 VAC models do not come with plugs) and are intended for indoor use in non-hazardous areas only.
Not CSA approved
Product Type Drum
Min temperature (º F) 50
Max temperature (º C) 232
Watts 1600
Diameter 22.3 in
Min temperature (º C) 10
Power (Amps) 6.6
Power (VAC) 240
Drum Size (gal.) 55
Max temperature (º F) 450
Drum Type Metal
Model FGDHC55240
Qty/ea 1
Brand BriskHeat

Accessories
Qty/Price
Availability

at $99.00 USD/EACH

Usually ships in 14 days.




Technical Resources
Virtual Catalog (PDFs)
  • Catalog - English
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
    Questions and Answers
    Product Reviews