United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
6 >
Representative photo only
5 To 5 PSId Handheld Differential Pressure Gauges
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-68600-06
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17090-31
NIST-Traceable Calibration; Manometers
$140.00 USD/EACH/EACH
Usually ships in 4 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pressure & Vacuum Instruments
>
Digital and Analog Manometers
>
Handheld Differential Pressure Gauge
Share
|
-5 To 5 PSId Handheld Differential Pressure Gauges
EW-68600-06
$649.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Intrinsically safe models are ideal for refineries, petrochemical plants, and mines - certified EEx ia IIC T4
Select one of nine units
Features zero, hold, max., min., out of range, and low battery functions
Smooth fluctuating readings with filter functions
Displays average of sixteen pressure readings
What's included:
one 9 V battery, two 2-ft lengths of nylon tubing. Intrinsically safe models also include an allen wrench (for battery replacement), and a hard plastic carrying case.
Specifications
Product Type
Manometers
Media compatibility
clean, noncorrosive, inert gases
Accuracy
±0.2% full scale
Unit of Measurement
psi, " H
2
O, cm H
2
O, " Hg, mm Hg, mbar, torr, kPA, and kg/cm
2
Resolution
0.001 psi
Range
-5 to 5 psid
Process connection
1/8" BSP
Battery life
90 hours, average
Power
9 V battery (included)
CE Compliance
Yes
Display
4-digit LCD, 7/16"H
Accessories
Qty
/
Price
Availability
Batteries, 9 V, 4/pack
EW-09376-04
at
$25.60 USD/PACKAGE of 4
In stock.
Add to Cart
PVC tubing, 4 mm ID x 6 mm OD, 25 ft/pack
EW-95616-00
at
$100.00 USD/CASE
Usually ships in 15 days.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Manometers
EW-17090-31
at $140.00 USD/EACH
Usually ships in 4 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Anemometers
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Show All...
Pressure Unit Conversion Constants
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title