United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
2
EW-91100-63
60 Piece Hot Box Insulated Tool Kit
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-91100-63
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Industrial Equipment
>
Tools
>
Tool Kits
>
Insulated Tool Kits
Share
|
60 Piece Hot Box Insulated Tool Kit
EW-91100-63
$3,075.00 USD/EACH
Usually ships in 4 days.
Qty
:
Add to Cart
Save to shopping list
Helps workers meet OSHA requirements
Rated for exposure up to 1000VAC and dielectrically tested at 10,000VAC
Meet or exceed ASTM 1505-07 and IEC 900 Standards for Insulated Hand Tools
Helps comply with OSHA 1910.333 (c)(2), and NFPA 70E 2004
These quality insulated tools are made with impact-resistant and flame-resistant material. Two-color insulation makes inspection easier which adds to the overall safety. If the yellow under layer is showing, the tool may no longer be insluated properly and should be considered for replacement and shall be removed from service.
Custom kits and individual tools available. Call our Application Specialists for more information.
60 Piece Hot Box Insulated Tool Kit Includes:
5 slotted screwdrivers: 1/8 x 2'', 3/16 x 4'', 3/16 x 6'', 1/4 x 6'', 5/16 x 6''
2 Phillips screwdrivers: #2 x 4'', #2 x 8''
7 6'' long nutdrivers: 3/16'', 1/4'', 5/16'', 11/32'', 3/8'', 7/16'', 1/2''
Torque wrench 30-150 in/lb
3/8'' drive reversible ratchet
3'' and 6'' extensions, 3/8'' drive
10 pcs. 5/16'' to 7/8'', 3/8'' dr socket set
11 pcs. 3/8'' to 1'' box end wrench set
14 pcs. 1/16'' to 1/2'', 6'' long arm, type hex wrench set
7 1/2'' diagonal cutting plier
8'' needle nose plier
9'' Linesman's plier
9 1/2'' cable cutting plier
10'' water pump plier
Combination stripper/crimper plier
Plastic box, including tool rolls
Specifications
Product Type
Tool Sets
Qty/pk
60
Manufacturer number
TK60
Brand
Salisbury
Technical Resources
Related technical articles (Roll over for summary)
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
ABCs of DMMs
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title