log in or register
60 mL B-D plastic disposable syringe, slip tip, 40 / box
EW-07940-28
$51.00 USD/BOX
Call 1-888-358-4705 for availability.
Qty
  • Ideal for use with syringe filters
Use these disposable, sterile, polypropylene syringes for general-purpose applications. Syringes come in an easy-to-open sealed package and can be autoclaved out of the package. Needles are not included; order separately.
Material Polycarbonate
Product Type Chromatography syringe
Capacity (mL) 60
Syringe type Luer-Loc
Graduations (mL) 1 mL, 2 oz in 1/4 oz
Sterile Yes
Quantity 40/bx
Brand B-D
Manufacturer number 14823 43
Accessories
Qty/Price
Availability
at $12.50 USD/BOX of 100
Call 1-888-358-4705 for availability.
at $12.50 USD/BOX of 100
Usually ships in 14 days.
at $12.50 USD/BOX of 100
Usually ships in 12 days.
at $12.50 USD/BOX of 100
Usually ships in 14 days.
at $12.50 USD/BOX of 100
Call 1-888-358-4705 for availability.
at $12.50 USD/BOX of 100
Usually ships in 12 days.
at $14.00 USD/BOX of 100
Usually ships in 12 days.
at $14.00 USD/BOX of 100
Usually ships in 12 days.
at $14.00 USD/BOX of 100
Call 1-888-358-4705 for availability.
at $14.00 USD/BOX of 100
Usually ships in 15 days.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title