United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
29 >
Representative photo only
700P01 Pressure Module 10inH2O Diff Differential Pressure
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-68450-39
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17090-26
NIST-Traceable Calibration; Pressure Modules
$175.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pressure & Vacuum Instruments
>
Calibrators
>
Fluke Pressure Modules
Share
|
700P01 : Pressure Module 10inH2O Diff Differential Pressure
EW-68450-39
$1,299.95 USD/EACH
Usually ships in 14 days.
Qty
:
Add to Cart
Save to shopping list
Pressure readings update twice per second, and can be displayed in up to 11 different units
Fluke offers 29 Pressure Modules for the Fluke 741B, 743B, as well as 744 & 754 Documenting Process Calibrators, the Fluke 717 and 718 Pressure Calibrators, plus the 725 Multifunction Process Calibrator. Gauge, differential, dual range, absolute and vacuum modules are available.
Available for sale in the United States and Canada only.
Specifications
Product Type
Pressure/Vacuum Calibrators
Brand
Fluke
Manufacturer number
FLUKE-700P01
Model
FLUKE-700P01
Make
PROCESS TOOLS
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Pressure Modules
EW-17090-26
at $175.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Pressure Unit Conversion Constants
Show All...
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title