log in or register
710000000021:1CH.Chart Rcdr Mrc7000 Circular Chart Recorder Standard Cover 115V Ac
EW-49001-88
$1,002.25 USD/EACH
Call 1-888-358-4705 for availability.
Qty
    A general purpose, microcontroller based, one or two pen, 10" circular chart recorder available in four versions; basic recorder, limit recorder, recording controller, and recording profile controller. The controller is capable of controlling any process from simple on/off to dual 4-20mA outputs with full PID on both pens. Can be panel or surface mounted.
    Standard features
    • 4 Digit display per pen
    • Universal Isolated sensor input: thermocouple, RTD, mA, mV, or V
    • Chart speed from 0.1 to 999.9 hours
    • Disposable fiber tipped pens: red, green
    Optional capabilities
    • 2 Programmable alarms per pen
    • Up to 8 relay type outputs
    • Up to four 4-20mA outputs
    • Process value retransmission
    • RS-485 Communications
    • 24V Transmitter power supply
    • Door lock
    • Water-tight enclosure
    Optional controller capabilities
    • On-off, time proportioning, or current proportioning control
    • Single or dual control output
    • Position proportioning for use with slidewire feedback
    • Relay, SSRD, or mA outputs
    • Remote setpoint input
    Optional profiler capabilities
    • Profile one or two pens
    • 8 Profiles, each with 6 ramps and 6 soaks
    • Up to 6 event relay outputs
    • Remote Run/Hold
    Product Type Chart Recorder/Controllers
    Model 710000000021
    Manufacturer number 710000000021
    Power 115 VAC
    Brand Partlow
    Accessories
    Qty/Price
    Availability
    at $19.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $19.00 USD/EACH
    In stock.
    at $21.00 USD/EACH
    Call 1-888-358-4705 for availability.
    at $21.00 USD/EACH
    Usually ships in 14 days.
    at $21.00 USD/EACH
    In stock.
    at $19.00 USD/EACH
    Usually ships in 9 days or sooner.
    at $21.00 USD/EACH
    In stock.
    at $19.00 USD/EACH
    Call 1-888-358-4705 for availability.
    Box Of 100 12inCHRTS
    EW-49003-05
    at $21.00 USD/EACH
    In stock.
    For All Elec Rec
    EW-49004-90
    at $7.00 USD/EACH
    In stock.
    at $35.00 USD/PACKAGE
    In stock.
    Pens: Green 5/pk
    EW-49004-99
    at $35.00 USD/PACKAGE
    In stock.
    Model:64402001
    EW-49005-23
    at $17.00 USD/EACH
    Usually ships in 14 days.
    Calibrations
    Qty/Price
    Availability
    at $210.00 USD/EACH
    Usually ships in 14 days.
    Technical Resources
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title