Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

8 Port Clear Drying Chambers

EW-03337-66
$1,990.00 USD/EACH

Call 1-888-358-4705 for availability.
Qty
    Use acrylic chambers with aluminum tops to accommodate three-shelf sample stands (order separately) for bulk freeze drying process with visual monitoring. Select model 03337-66 for up to eight additional valved ports (e.g. for Fast-Freeze Flasks). Three-shelf stands are independently height-adjustable. Available for ambient temperature use (vials, serum bottles, and bulk samples) or heated (to 40° C, variable temperature controlled) to accelerate freeze drying of bulk samples.
    Product Type Drying Chambers
    Dimensions 12.25 in H x 14.625 in Dia.
    Manufacturer number 7443500

    Accessories
    Qty/Price
    Availability

    at $3,105.00 USD/EACH

    Call 1-888-358-4705 for availability.

    at $3,130.00 USD/EACH

    Call 1-888-358-4705 for availability.

    at $675.00 USD/EACH

    Call 1-888-358-4705 for availability.
    Build a complete system




    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title