Marathon BI-030014 8 x 40 Binoculars with Built-In Sun Filter
Representative image only.
Marathon Mfr # BI-030019 – Item # EW-46160-10
Beyond binoculars with built-in compass or sun filter
  • Ideal for spectator sports, environmental studies, and communication command activities
  • Bright, sharp images with glass coated designed lenses
  • Tripod mountable for hands-free observation
  • 5 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Magnification8x
  • Viewing Area1.6\" (40 mm) diameter
  • Mounting TypeTripod-mountable
  • Description8 x 40 Binoculars with Built-In Sun Filter
  • Warranty5 Years
5 Year Warranty
Key Features
  • Ideal for spectator sports, environmental studies, and communication command activities
  • Bright, sharp images with glass coated designed lenses
  • Tripod mountable for hands-free observation
  • 5 year warranty
More About this Item
Binoculars with compass model 46160-00
These high power binoculars with built-in 360° compass and reticles provide quick determination of distance and direction of objects. Waterproof and fog-proof design will withstand rain, fog, or high humidity conditions. Features 27.4-mm long eye-relief to reduce eye strain, twist-up eyecups, individual focus, and magenta coated lenses for superior image brightness.
What’s included: Nylon carrying case, neck strap, eye-cup and front lens protectors, tripod adapter socket, lens cloth and silicon gel.
Binoculars with sun filter model 46160-10
Designed with the outdoors enthusiasts in mind, these bright yellow binoculars offer sun filters to help protect against glare during viewing. Ergonomic rubber armor housing stands up to rugged fie