log in or register
96-Well Equilibrium Dialyzer - 5,000 Daltons
EW-02896-70
$430.00 USD/EACH
Usually ships in 4 days.
Qty
  • Lets you simultaneously assay up to 96 samples
  • Each well contains a separate membrane to eliminate cross-contamination
  • High reproducibility from well to well
The single-use equilibrium dialyzer is a novel product for the simultaneous assay of 96 samples. Each well has a separate membrane to eliminate cross contamination. The high-quality regenerated cellulose membranes are free of sulfur and heavy metal contamination.
Wells are sealed with 8-cap strips, so either a row of wells or all 96 wells can be used depending on the experiment. Reproducibility is very high across the different wells, and sample recovery is above 95%.
What’s included: twelve 8-cap top strips and twelve 8-cap bottom strips to seal wells. Rotator is required; order separately.
Product Type Dialyzer
Material Regenerated Cellulose
Membrane MWCO (Daltons) 5000
Manufacturer number 74-2330
Accessories
Qty/Price
Availability
at $755.00 USD/EACH
Usually ships in 7 days.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title