United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 7
of
9 >
Representative photo only
AIM600 Programmable Block Digestion System 28 Place With 250 mL Tubes
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-78871-51
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Shop by Industry
>
Chemical Property Testing
>
Fat, Protein, Alcohol & Crude Fiber Analysis
>
AIM600 Programmable Digestion Systems
Share
|
AIM600 Programmable Block Digestion System, 28 Place With 250 mL Tubes
EW-78871-51
$8,360.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Increase productivity with programmable digestions and large vial capacity
Perform digestions of aqueous samples for metals analysis by Atomic Absorption (AA) or Inductively Coupled Plasma (ICP)
Conduct Kjeldahl digestion of food products or plant tissue for nitrogen & phosphorous analysis
Free up valuable time by digesting large numbers of samplesup to 50 vialssimultaneously
Eliminate the need for repetitive small batch digestions with these compact block digestion systems. These systems can handle sample batches of 28 or 50 at a time. Run your digest overnight and have a full batch of digested samples ready for analysis at the start of the next day. The temperature of each digestion position is tightly regulated and controlled, ensuring that all the samples in each batch are digested equally. Store up to nine different programs, each with up to forty steps, for automating the most complex digestion procedures, allowing reproducibility from batch to batch, operator to operator. Controlled heating rates, as low as 1°C per minute, protect against the bumping and splattering of reactive samples.
A built in safety cut-out feature within the controller is triggered automatically if the programmed temperature is exceeded by 30°C or if the maximum set temperature is exceeded. The controller is separate from the digestion block, protecting the electronics and the operator from any digest acid fumes. Apart from the power and reagents, these block digestion systems have no ongoing
consumable costs and do not use any limited life components.
What's Included:
microprocessor controller, digestion block, draft shield, set of digestion tubes, and 9-ft power cord with plug.
Specifications
Product Type
Programmable Block Digestion System
Set-point accuracy
±2°C
Max operating temperature (º F)
842
Max operating temperature (º C)
450
Dimensions (" H)
5 3/4
Dimensions (" W)
23 1/2
Dimensions (" D)
11
Power
Block: 220 to 260 VAC, 50/60 Hz; Controller: 100 to 260 VAC, 50/60 Hz
Accessories
Qty
/
Price
Availability
Tubes, Straight, 75 mL, 50/pk
EW-78870-70
at
$819.00 USD/PACKAGE
Usually ships in 2 days.
Add to Cart
Tubes, Volumetric, 75 mL, 50/pk
EW-78870-72
at
$1,026.00 USD/PACKAGE
Usually ships in 13 days.
Add to Cart
Tubes, Volumetric, 100 mL, 50/pk
EW-78870-76
at
$1,036.00 USD/PACKAGE
Usually ships in 2 days.
Add to Cart
Tubes, Straight, 250 mL, 28/pk
EW-78870-78
at
$794.00 USD/PACKAGE
Usually ships in 2 days.
Add to Cart
Tubes, Volumetric, 250 mL, 28/pk
EW-78870-80
at
$811.50 USD/PACKAGE
Usually ships in 2 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Electron zap turns flimsy plastic into sturdy shrink wrap
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title