United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
EW-81200-00
ATAGO
®
Semiautomatic Polarimeter
Click to enlarge
More Product Details
Specifications
Accessories
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-81200-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Material Testing & Physical Testing
>
Polarimeters
>
ATAGO Semiautomatic Polarimeter
Share
|
ATAGO Semiautomatic Polarimeter
EW-81200-00
$6,100.00 USD/EACH
Usually ships in 7 days.
Qty
:
Add to Cart
Save to shopping list
Measurement is digitally displayed as either angle of rotation or International Sugar Scale
Simple and precise push-button adjustment of visual fields
Zero indicator light confirms accurate calibration
This polarimeter features a switch-selectable digital readout that displays either the angle of rotation (AOR) or the International Sugar Scale (ISS). Polarimeter offers push-button rotation switches to adjust the left and right visual fields to an equal brightness—no more imprecise manual adjustments. To calibrate, simply zero the polarimeter using distilled water, and within moments a zero indicator light confirms successful calibration.
Depending on your accuracy needs, choose the fast mode or slow mode to adjust the visual field brightness. The fast mode allows you to quickly adjust the polarimeter for an approximate reading by continuously rotating the analyzer. The slow mode lets you accurately adjust the polarimeter by rotating the analyzer 0.05° at a time. Once visual field brightness is equal, the digital display will give you the reading.
The polarized light source is an LED with a filter for 589 nm. For best results, use the 200-mm tube for transparent liquids and the 100-mm tube for cloudy, impurity-filled samples. The tubes contain a bubble trap that prevents errors caused by air in your samples.
What's Included:
One each 100- and 200-mm tubes and a 7-ft (2.1 m) power cord with three-prong plug.
Specifications
Product Type
Semiautomatic Polarimeter
Range
Rotation: +180° to -179.95°
ISS:+130°Z to -130°Z
Resolution
Angle: 0.05°
ISS: 0.1°Z
Accuracy
Angle of rotation: ±0.1°
ISS: ±0.3°Z
Light source
LED with filter for 589 nm
Tube length
up to 200 mm
Dimensions
17"W x 11-13/16"H x 8-9/16"D (43.2 x 30 x 22 cm)
Power
100 to 240 VAC, 50/60 Hz
Accessories
Qty
/
Price
Availability
Replacement tube; 100 mm, 5 mL
EW-81200-50
at
$135.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Replacement tube; 200 mm, 9 mL
EW-81200-52
at
$135.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Related technical articles (Roll over for summary)
Principles of Polarimetry
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title