Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Acetate Faceshield, Clear, Anti Fog

EW-86478-17
$28.00 USD/EACH

  • The chemical protection of acetate at an economical price
Ratcheting head gear allows for fast and easy adjustment. Face shield curves naturally to fit facial contours ensuring reliable protection from splash, particles, and flying debris. Acetate window is chemical splash, and impact resistant. Window easily attaches with keyhole design and locks in the up or down position depending on need. Nonglare finish allows for high clarity.
Compliance: ANSI Z87.1-2003, CUL, CAN/CSA-Z94.3 Standards
Product Type Face Shields
Color Blue
Lens Clear
Coating Antifog
Compliance ANSI Z87.1-2003, CUL, CAN/CSA-Z94.3 Standards
Brand Sellstrom
Model 39140
Manufacturer number 39140

Replacement parts
Qty/Price
Availability

at $10.50 USD/EACH
In stock.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title