United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 8
of
8
EW-08900-64
Acrylic Desiccator with gas port
Click to enlarge
More Product Details
Specifications
Accessories
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-08900-64
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17030-28
NIST-Traceable Calibration; Standard Accuracy Humidity and Temperature
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Desiccators
>
Desiccator Cabinets
>
Acrylic Desiccators with Gas Ports
Share
|
Acrylic Desiccator with gas port; cabinet color, amber; exterior dimensions, 24"W x 24"H x 18"D
EW-08900-64
$1,235.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Easily monitor humidity levels with built-in hygrometer
Gas ports let you flush unit and maintain stable internal conditions
These durable desiccators have a temperature range of 40 to 120°F (4 to 49°C). They are made of heavy-duty, 1/4" thick acrylic. Closed cellular PVC gaskets prevent system leaks—gaskets will not take a set or keep an impression. Two gas ports (1/8" NPT x 1/4" hose barb inlet; 1/4" NPT x 1/4" hose barb outlet) are included. Not designed for vacuum use.
Choose clear units for easy viewing of contents, or choose amber units for protecting light-sensitive compounds from ultraviolet light. All models are easy to access and clean; slip-away hinges allow complete removal of door when needed. Adjust shelf positions to suit your needs. Models 08900-50 and -52 include two shelves; will hold five shelves. All other models include four shelves; will hold ten shelves.
Models 08900-58, -60, -62 and -64 must be shipped motor freight.
Specifications
Product Type
Cabinet Desiccator
Material
Acrylic
Color
amber
Exterior Dimensions (" W)
24
Exterior Dimensions (" H)
24
Exterior Dimensions (" D)
18
Brand
Plas-Labs
Accessories
Qty
/
Price
Availability
FEP Tubing, 1/8"ID x 3/16"OD, 25 ft/pack
EW-06406-64
at
$30.50 USD/PACKAGE
In stock.
Add to Cart
Shelf F/08900-64
EW-08900-85
at
$113.00 USD/EACH
Call 1-888-358-4705 for availability.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Standard Accuracy Humidity and Temperature
EW-17030-28
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Equipping a Laboratory: A Study in Customer Service Excellence
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title