United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
16 >
EW-11712-14
Adam Highland Portable Balance, 120g X 0.001g
Click to enlarge
More Product Details
Specifications
Accessories
Related products
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-11712-14
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-52
NIST Traceable Report With Data, Balances/Scales - all types
$140.00 USD/EACH/EACH
Usually ships in 7 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Toploading Portable Balances
>
Adam Equipment Portable Balances
>
Adam Equipment Highland Portable Balances
Share
|
Adam Highland Portable Balance, 120g X 0.001g
EW-11712-14
$425.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Loaded with features for everyday weighing applications
Internal rechargeable battery for convenient portable operation
Standard draft shield offers greater weighing accuracy
Unique shape allows convenient stackable storage of multiple balances
Loaded with features that include multiple weighing units, RS232 and USB interfaces, rechargeable battery pack, capacity tracker, security lock-down port, and below-balance weighing hook for density and specific gravity applications. A standard removable draft shield eliminates wind disturbances outdoors or in drafty environments. Backlit display with large 1/2''-high digits offers a clear view of weighing results.
What's Included:
AC power adapter, internal rechargeable battery, and 4" H x 5" dia (10.2 x 12.7 cm) draft shield
Specifications
Product Type
Portable Balances
Auto shutoff
Yes
Calibration
Internal
Capacity (g)
120
Capacity (oz)
4.2
Housing dimensions
17.1 x 7.6 x 25.4 cm
Dimensions (" W)
6-3/4
Dimensions (" H)
3
Dimensions (" D)
10
Display
LCD
Pan size (dia)
4-1/2" (11.4 cm)
Readability (g)
0.001
Linearity (g)
±0.003
Repeatability (g)
±0.003
Weighing units
g, ct, lb, oz, d, gn, ozt, dwt, momme, taels (3), Newtons
Stabilization time
2-4 seconds
Power (VAC)
115
Battery
rechargeable battery (included)
Draft shield (WxHxD)
130mm dia. X 100mm high
Stability Indicator
Yes
Output
RS-232
Brand
Adam Equipment
Model
HCB123
Accessories
Qty
/
Price
Availability
Thermal Printer for Balances with an RS-232 Interface
EW-10000-38
at
$395.00 USD/EACH
Usually ships in 10 days.
Add to Cart
Replacement Paper for Thermal Printer 10000-38 (pack of 5)
EW-10000-39
at
$65.00 USD/PACKAGE
Usually ships in 10 days.
Add to Cart
ATP Adam Thermal Printer
EW-11120-78
at
$335.00 USD/EACH
Usually ships in 6 days.
Add to Cart
Thermal Paper For Adam ATP (10 pack)
EW-11120-79
at
$90.00 USD/PACKAGE
Usually ships in 6 days.
Add to Cart
In use wet cover for HCB
EW-11120-83
at
$20.00 USD/EACH
Usually ships in 6 days.
Add to Cart
RS232 cable
EW-11120-93
at
$35.00 USD/EACH
Usually ships in 6 days.
Add to Cart
USB cable
EW-11120-94
at
$35.00 USD/EACH
Usually ships in 6 days.
Add to Cart
RS232 to USB Cable
EW-11120-95
at
$50.00 USD/EACH
Usually ships in 6 days.
Add to Cart
Adam DU Software
EW-11120-96
at
$95.00 USD/EACH
Usually ships in 6 days.
Add to Cart
Related products
Qty
/
Price
Availability
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
EW-01019-11
at $43.00 USD/EACH
In stock.
Add to Cart
Calibrations
Qty
/
Price
Availability
NIST Traceable Report With Data, Balances/Scales - all types
EW-17101-52
at $140.00 USD/EACH
Usually ships in 7 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Weighing Unit Conversion Table
Common Balance Terms
Equipping a Laboratory: A Study in Customer Service Excellence
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title