log in or register
Adam PGW Precision Toploading Balance, 450 g x 0.001 g - 115 VAC
EW-11701-41
$1,075.00 USD/EACH
Usually ships in 14 days.
Qty
  • Improved response times and enhanced stability and features
  • GLP-compliant printouts via RS-232 connection
  • Digital filters compensate for vibration, drafts or temperature changes
  • Parts counting, check weighing, percent weighing, animal weighing
  • Below-balance weighing hook
  • Date and time stored in memory
  • Oversized backlit LCD
These balances feature multiple weighing modes including parts counting, check weighing, animal weighing, net/total formulation, liquid density, solid density, and percentage weighing. An oversized backlit LCD with split segments and 1” high digits offers easy visibility. The capacity tracker graphically displays percentages and notifies you of any approaching overloads. All models feature an RS-232 port for configuration to a printer—providing GLP-compliant reports after each calibration.
A second output port may be linked to other devices such as USB, Ethernet, or a remote display whenever additional communication outputs are desired. The balances feature 17 weighing units including one custom unit and seven weighing modes. Toploading balances offer choice of internal or external calibration; deactivate internal calibration at any time to perform external calibrations.
What's Included:
AC adapter with 6-ft cord, below-balance weighing hook, and lock-down Kensington™ type mounting point
Product Type Toploading Balances
Calibration External
Capacity (g) 450
Capacity (oz) 16
Readability (g) 0.001
Repeatability (g) 0.001
Power (VAC) 115
Brand Adam Equipment
Model PGW453E
Accessories
Qty/Price
Availability
at $375.00 USD/EACH
Usually ships in 10 days.
at $25.00 USD/EACH
Usually ships in 10 days.
at $395.00 USD/EACH
Usually ships in 10 days.
at $65.00 USD/PACKAGE
Usually ships in 10 days.
at $20.00 USD/EACH
Usually ships in 10 days.
at $45.00 USD/EACH
Usually ships in 10 days.
at $335.00 USD/EACH
Usually ships in 6 days.
at $90.00 USD/PACKAGE
Usually ships in 6 days.
RS232 cable
EW-11120-93
at $35.00 USD/EACH
Usually ships in 6 days.
RS232 to USB Cable
EW-11120-95
at $50.00 USD/EACH
Usually ships in 6 days.
Adam DU Software
EW-11120-96
at $95.00 USD/EACH
Usually ships in 6 days.
at $241.00 USD/EACH
Call 1-888-358-4705 for availability.
Related products
Qty/Price
Availability
at $55.00 USD/PACKAGE of 500
In stock.
at $43.00 USD/EACH
In stock.
Calibrations
Qty/Price
Availability
at $140.00 USD/EACH
Usually ships in 7 days.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title