log in or register
Adam PW Analytical Balance, 250 g x 0.0001 g, 220VAC
EW-11700-58
$2,045.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
  • Improved response times and enhanced stability and features
  • GLP-compliant print outs via RS232 connection
  • Digital filters compensate for vibration, drafts or temperature changes
  • Parts counting, check weighing, percent weighing, animal weighing
  • Below balance weighing hook
  • Date and time stored in memory
  • Oversized backlit LCD
These balances feature multiple weighing modes; including parts counting, check weighing, animal weighing, net/total formulation, liquid density, solid density, and percentage weighing. An oversized backlit LCD with split segments and 1" high digits offers easy visibility. The capacity tracker graphically displays percentages and notifies the user of any approaching overloads. All models feature an RS-232 port for configuration to a printer—providing GLP-compliant reports after each calibration.
A second output port may be linked to other devices such as USB, Ethernet, or a remote display whenever additional communication outputs are desired. The balances feature 17 weighing units including one custom unit and seven weighing modes. Toploading balances offer choice of internal or external calibration; deactivate internal calibration at any time to perform external calibrations.
What's Included:
AC adapter with 6-ft cord, below balance weighing hook, and lock-down Kensington type mounting point
Product Type Analytical Balances
Auto shutoff digital filters and suppression of the last digit on the display
Calibration Internal
Capacity (g) 250
Capacity (oz) 9
Readability (g) 0.0001
Linearity (g) 0.0002
Repeatability (g) 0.0002
Weighing units g, ct, mg, gn, oz, dwt, mo.
Output RS-232
Brand Adam Equipment
Model PW254_220V
Accessories
Qty/Price
Availability
at $375.00 USD/EACH
Usually ships in 10 days.
at $25.00 USD/EACH
Usually ships in 10 days.
at $395.00 USD/EACH
Usually ships in 10 days.
at $65.00 USD/PACKAGE
Usually ships in 10 days.
at $20.00 USD/EACH
Usually ships in 10 days.
at $45.00 USD/EACH
Usually ships in 10 days.
at $335.00 USD/EACH
Usually ships in 6 days.
at $90.00 USD/PACKAGE
Usually ships in 6 days.
Anti-vibration Table
EW-11120-92
at $675.00 USD/EACH
Usually ships in 6 days.
RS232 cable
EW-11120-93
at $35.00 USD/EACH
Usually ships in 6 days.
RS232 to USB Cable
EW-11120-95
at $50.00 USD/EACH
Usually ships in 6 days.
Adam DU Software
EW-11120-96
at $95.00 USD/EACH
Usually ships in 6 days.
Related products
Qty/Price
Availability
at $43.00 USD/EACH
In stock.
Calibrations
Qty/Price
Availability
at $140.00 USD/EACH
Usually ships in 7 days.
Technical Resources
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title