United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 6
of
12 >
Representative photo only
Adam WBZ 30a Price Computing Scale 30lb Capacity 0 01lb Readability 220V
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-11810-39
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Price Computing and Shipping Scales
>
Adam Equipment WBZ Series Wash Down Scales
Share
|
Adam WBZ 30a Price Computing Scale, 30lb Capacity, 0.01lb Readability ,220V
EW-11810-39
$355.00 USD/EACH
Usually ships in 6 days.
Qty
:
Add to Cart
Save to shopping list
The Adam Equipment WBZ wash down approved retail scales are ideal for food preparation, packaging plants, agricultural facilities, and other retail applications.
IP66 rated for wash down
Large backlit LCD on both front and back panels
Displays weight, unit price, and total price
Price displayed in multiple currency
Rechargeable battery; up to 90 hours operation
Auto sleep / power down function to save battery life
Stainless steel pan
Full color coded, sealed keypad with numeric entry
Level indicator with adjustable levelling feet
Non-slip adjustable levelling feet
Full range tare
Precision load cell technology
Low battery indication
Lightweight and portable
Large stainless steel pan
Zero Tracking
Simple user-friendly operation
Tare function
The WBZ scales feature an IP66 rated wash down housing with a large stainless steel weighing pan. The larger backlit display shows weight, price per unit, and an easy to read extra large total cost display. They operate with an AC adapter or rechargeable battery pack for portability; both are included.
What's Included:
Internal rechargeable battery pack and AC adapter.
Specifications
Product Type
Price Computing Scales
Auto shutoff
Yes
Calibration
External
Capacity (lbs)
30
Dimensions (" W)
10
Dimensions (" H)
6
Dimensions (" D)
9
Display
Backlit LCD
Platform dimensions
8.3" x 6.8"
Readability (lbs)
0.01
Weighing units
lb, kg
Stabilization time
2 sec
Power (VAC)
220
Battery
Rechargeable
Low battery indicator
Yes
RS-232 interface
Yes
Brand
Adam Equipment
Model
WBZ 30a -220V
Technical Resources
Related technical articles (Roll over for summary)
Weighing Unit Conversion Table
Common Balance Terms
Equipping a Laboratory: A Study in Customer Service Excellence
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title