dummy
EW-07902-26
- SafetyPrime valve recirculates reagent during priming to reduce spurting and conserve reagent.
- Safety discharge system reduces the risk of accidental dispensing.
- Valve block rotates 360° so that the volume display and bottle label are always visible.
- Floating piston design for smooth, one-handed operation
- Reinforced discharge tube with a threaded safety cap
- Polypropylene cylinder sleeve helps protect the user in case of accidental breakage.
- Digital models feature Easy Calibration for quick adjustments in the labwithout having to retest.
- Can be calibrated for ISO/GLP compliance
- Fit directly onto 45-mm screw-neck bottles. Come with polypropylene adapters for other neck sizes. Order bottles separately).
Dispensette® III Dispensers Safely dispense bases, acids, aqueous solutions, hydrochloride salts, and many organic solvents*. These dispensers are available with fixed or adjustable volume.
DISPENSETTE—Reg TM BRAND GMBH & CO.
DRIERITE—Reg TM W.A. Hammond Drierite Co.
PYREX—Reg TM Corning Glass Works
PTFE, TEFZEL—Reg TM E.I. DuPont de Nemours & Co.
| Product Type |
Dispenser |
| Volume (mL) |
100 |
| Accuracy |
±0.5% |
| Division (mL) |
1 |
| Range (mL) |
10 - 100 |
| Reproducibility |
≤0.1% |
| Wetted parts |
Platinum-iridium valve spring, borosilicate glass valve balls and cylinder, PTFE washers, PFa-coated piston, ETFE valve bodies and valve block, AlO3 valve seats, and PP safety cap. |
| Includes |
PP adapters for threaded bottle neck size 28, 32, & 38 mm Telescoping filling tube (125 to 240 mm) |
| Autoclavable |
Yes |
| Brand |
Dispensette |
| Manufacturer number |
4701171 |
| Model |
4701171 |
at $206.00 USD/CARTON
In stock.
at $38.50 USD/EACH
Usually ships in 11 days.
at $104.00 USD/EACH
Usually ships in 3 days.
at $123.00 USD/EACH
Call 1-888-358-4705 for availability.
at $23.75 USD/EACH
Call 1-888-358-4705 for availability.
at $24.75 USD/EACH
Call 1-888-358-4705 for availability.
at $25.00 USD/EACH
Call 1-888-358-4705 for availability.
at $66.00 USD/EACH
Usually ships in 9 days.
Technical Resources
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334