Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Adjustable-Volume analog dispenser - 1-10 mL

EW-07902-18
$434.00 USD/EACH

  • SafetyPrime valve recirculates reagent during priming to reduce spurting and conserve reagent.
  • Safety discharge system reduces the risk of accidental dispensing.
  • Valve block rotates 360° so that the volume display and bottle label are always visible.
  • Floating piston design for smooth, one-handed operation
  • Reinforced discharge tube with a threaded safety cap
  • Polypropylene cylinder sleeve helps protect the user in case of accidental breakage.
  • Digital models feature Easy Calibration for quick adjustments in the lab—without having to retest.
  • Can be calibrated for ISO/GLP compliance
  • Fit directly onto 45-mm screw-neck bottles. Come with polypropylene adapters for other neck sizes. Order bottles separately).
Dispensette® III Dispensers Safely dispense bases, acids, aqueous solutions, hydrochloride salts, and many organic solvents*. These dispensers are available with fixed or adjustable volume.
DISPENSETTE—Reg TM BRAND GMBH & CO.
DRIERITE—Reg TM W.A. Hammond Drierite Co.
PYREX—Reg TM Corning Glass Works
PTFE, TEFZEL—Reg TM E.I. DuPont de Nemours & Co.
Product Type Dispenser
Volume (mL) 10
Accuracy ±0.5%
Division (mL) 0.2
Range (mL) 1 - 10
Reproducibility ≤0.1%
Wetted parts Platinum-iridium valve spring, borosilicate glass valve balls and cylinder, PTFE washers, PFa-coated piston, ETFE valve bodies and valve block, AlO3 valve seats, and PP safety cap.
Includes PP adapters for threaded bottle neck size 24, 28, 32, & 38 mm
Telescoping filling tube (125 to 240 mm)
Autoclavable Yes
Brand Dispensette
Manufacturer number 4701141
Model 4701141

Accessories
Qty/Price
Availability

at $206.00 USD/CARTON

In stock.

at $38.50 USD/EACH

Usually ships in 11 days.

at $104.00 USD/EACH

Usually ships in 3 days.

at $123.00 USD/EACH

Call 1-888-358-4705 for availability.

at $23.75 USD/EACH

Call 1-888-358-4705 for availability.

at $24.75 USD/EACH

Call 1-888-358-4705 for availability.

at $25.00 USD/EACH

Call 1-888-358-4705 for availability.
Calibrations
Qty/Price
Availability

at $66.00 USD/EACH
Usually ships in 9 days.




Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title