log in or register
Adjustable leg workstation, ESD laminate surface, 30" x 60"
EW-47545-46
$595.00 USD/EACH
Usually ships in 7 days.
Qty
  • Adjustable height delivers flexability and ergonomics; accessories deliver productivity
  • Choose from basic and ESD workstations
  • Two styles are available: hydraulic hand crank and leg adjustable
  • All accessories can be used for either style workstation
  • Ships in 5 days!
Increase your productivity with these top-quality workstations. Choose from basic or ESD laminate, and hydraulic hand crank or adjustable leg. Worksurface laminate and industrial-grade particleboard is 1 ¼” thick, and has 180° rolled front edges. The worksurface is impact-resistant, and the ESD workstation is supplied with an ESD grounding kit. For workstations, see table below. The leg adjustable workstation comes in three sizes (see below) and worksurface height adjusts from 30” to 36” high in 2” increments. Adjustable Leg Extender Kit included, and leg extensions accepts floor glides. Heavy gauge support beams allow 1000 pounds distributed weight. The hydraulic hand crank workstation features cantilever leg design for unobstructed movement and plenty of legroom. All-welded heavy gauge steel supports up to 600 pounds static weight. The hand crank allows users to quickly and easily adjust the height of the worksurface a full 12”, from 30” to 42”, or any height in between! Choose between basic accessories such as shelves and drawers or Multi-Task System accessories such as tool trolley, multi bin panel, or CRT platform. MTS makes a great addition to existing workbenches-upgrade! Accessories even include air system for pneumatic tools. Accessories can be used for both styles of workstation, no drilling necessary, and are all listed below. Accessories can be used to upgrade your current workstations too!
Product Type Laboratory Worktables
Dimensions (" W) 60
Dimensions (" L) 30
Brand IAC
Manufacturer number QS-1006002-D
Model QS-1006002-D
Accessories
Qty/Price
Availability
at $254.00 USD/EACH
Usually ships in 7 days.
at $327.00 USD/EACH
Usually ships in 11 days.
at $363.00 USD/EACH
Call 1-888-358-4705 for availability.
at $161.00 USD/EACH
Usually ships in 12 days.
at $228.00 USD/EACH
Usually ships in 12 days.
at $198.00 USD/EACH
Usually ships in 12 days.
at $134.00 USD/EACH
Call 1-888-358-4705 for availability.
at $151.00 USD/EACH
Call 1-888-358-4705 for availability.
at $110.00 USD/EACH
Usually ships in 10 days.
at $128.00 USD/EACH
Call 1-888-358-4705 for availability.
Footrest tube, 60"W
EW-47545-74
at $65.00 USD/EACH
Usually ships in 10 days.
Footrest tube, 72"W
EW-47545-76
at $67.00 USD/EACH
Usually ships in 7 days.
at $72.00 USD/EACH
Usually ships in 7 days.
at $178.00 USD/EACH
Call 1-888-358-4705 for availability.
at $189.00 USD/EACH
Usually ships in 7 days.
at $83.00 USD/EACH
Call 1-888-358-4705 for availability.
at $90.00 USD/EACH
Usually ships in 7 days.
at $97.00 USD/EACH
Call 1-888-358-4705 for availability.
at $106.00 USD/EACH
Usually ships in 7 days.
Light fixture, 60"
EW-47546-00
at $232.00 USD/EACH
Usually ships in 7 days.
Light fixture, 72"
EW-47546-02
at $241.00 USD/EACH
Usually ships in 7 days.
at $70.00 USD/EACH
Usually ships in 7 days.
at $74.00 USD/EACH
Usually ships in 7 days.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title