Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Agar 500g

EW-89000-20
$95.00 USD/EACH

quantity discounts Quantity Discounts
Order Discount Price
[X]
6 10% $85.50/EACH
12 15% $80.75/EACH
24 20% $76.00/EACH
Usually ships in 7 days.
Qty

This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
Product Type Agar
Purity grade Purified
CAS Number 9002-18-0
Form Powder
Formula weight N/A
Synonyms Agar-Agar
Packaging Poly Bottle
Storage symbol Green
UN number N/A
Quantity (g) 500
DOT description Agar Not Regulated
Brand Cole-Parmer
Manufacturer number 900020
Quantity 500 g





Technical Resources
Certificate of Analysis
Enter lot number

or click to view a Sample C of A
 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title