United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
3
EW-33556-70
Representative Image. Check individual details.
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-33556-70
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Furniture & Fume Hoods
>
Fume Hoods
>
Fume Hood Accessories
>
Airflow Monitors
>
Wide-range Airflow Monitors for Fume Hoods
Share
|
Air Flow Monitor for Bio Safety Cabinets, 30 to 400 FPM Monitoring Range
EW-33556-70
$377.00 USD/EACH
Usually ships in 8 days.
Qty
:
Add to Cart
Save to shopping list
Reliable, durable sensor element
Simple field calibration
Multiple alarms—audible, visual, and relay
Monitors maintain accurate response even when using low-flow hood conditions. Alarms activate based on sustained low-flow condition. Use pushbuttons to set alarm and perform easy two-point calibration. Monitors will also indicate high sash condition (requires proximity switch, not included) with configurable time delay. Record hood performance using RS-232 connection to computer.
Model 33556-50 features a horn alarm with green and red LED indicators. The graphic display model 33556-60 comes with an external velocity meter and shows current velocity in fpm with bar graph depiction of recent hood airflow performance; calibration instructions appear on screen. Choose model 33556-70 for its extra wide measurement range. It includes a remote velocity probe which can be installed in your ductwork for use with biosafety cabinets.
What's Included:
low voltage transformer and mounting bracket for surface mount. Call our Application Specialists if a flush mount requiring a 2" x 3" (5.1 x 7.6 cm) opening is preferred.
Specifications
Product Type
Fume Hood Accessories
Power (VAC)
120
Power (Hz)
60
Brand
TEL UK
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
To B2 or not to B2- a Biological Safety Cabinet Story
Importance of Airflow Monitors for Fume Hoods
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title