log in or register
Air eliminator; 3/4" NPT(F) x 3/8" NPT(F)
EW-07600-92
$208.00 USD/EACH
Usually ships in 6 days.
Qty
  • Improve the circulation of pressurized liquids by eliminating air and other non-condensable gases
  • Mount at high points to provide immediate removal of air and gases
  • EPDM seals ensure positive shut-off
  • Cast-iron body with stainless steel internal components—can be serviced without disturbing the inlet piping connection
Typical applications: Use for both hot and cold liquid services
  • Cold water lines
  • Suction lines to pumps
  • Mixing tanks
  • Condensate return lines
  • Cool water lines on air compressors
  • Water storage tanks
Product Type Steam Handling fittings
Material Cast iron
Max temperature (º F) 450
Max temperature (º C) 232
Brand Spirax Sarco
Model 13WS.75 in 69843 
Manufacturer number 13WS 3/4" 69843 
Accessories
Qty/Price
Availability
at $16.00 USD/EACH
In stock.
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title