United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 7
of
8 >
EW-74440-60
Representative Image. Check individual details.
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-74440-60
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Positive Displacement Pumps
>
Air-Driven Pumps
>
Air-Powered Double-Diaphragm Pumps
>
Air-Operated Double Diaphragm Pumps
Share
|
Air-operated Diaphragm Pump, FDA-Compliant, 1-1/2" Triclamp, 106gpm
EW-74440-60
$3,121.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Complies for applications in food processing, pharmaceutical, and cosmetic applications.
Handle abrasive and shear-sensitive materials
Self-priming up to 24 ft of water
Deadhead against closed discharge without damage
Air distribution system can be serviced external to the pump allowing for true in-line maintenance. The air valve is guaranteed not to stall.
USDA AODD's wetted materials are electropolished 316 and 302/304 stainless steel. Non-wetted materials are electroless nickel-plated aluminum and polypropylene. FDA AODD's wetted materials are FDA-compliant stainless steel. All models pumps are fitted with white food-grade nitrile elastomers.
Specifications
Product Type
Air-Powered Diaphragm Pumps
Wetted materials
Stainless steel body, food-grade Santoprene® diaphragms, check valves, & valve seats
Port size
2" Tri-Clamp
Max flow rate (LPM)
401.3
Max viscosity (cp)
11,000
Max pressure
100 psi
Min temperature (º C)
-23
Max temperature (º C)
88
Min temperature (º F)
-10
Max temperature (º F)
190
Run dry
Yes
Suction lift (wet)
24 ft
Air inlet (SCFM)
10 to 55 scfm
Air inlet (connection)
3/4" NPT(F)
Air inlet (max psi)
100
Duty cycle
Continuous
Dimensions
16-3/4"L x 12-3/8"W x 21-7/8"H
Brand
Warren Rupp
Manufacturer number
T15B1SDSWTS600
Model
T15B1SDSWTS000
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Positive Displacement Pump Types
How to Read a Pump Curve
Guide to Liquid Pumps
Show All...
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title