United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
6 >
Representative photo only
Air operated double diaphragm drum pump polypropylene with PTFE diaphragm 12 GPM
Click to enlarge
More Product Details
Specifications
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-74024-00
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pumps
>
Positive Displacement Pumps
>
Air-Driven Pumps
>
Air-Powered Double-Diaphragm Pumps
>
Air-Operated Double-Diaphragm Drum Pumps
Share
|
Air-operated double-diaphragm drum pump; polypropylene with PTFE diaphragm, 12 GPM
EW-74024-00
$615.00 USD/EACH
Usually ships in 9 days.
Qty
:
Add to Cart
Save to shopping list
Safe and economical liquid transfer for manufacturing and water treatment environments
No sliding parts—pumps can run dry without damage
Variable flow rate and discharge pressure—one pump works for many applications
Portable and simple to install
Air-operated double-diaphragm drum pumps incorporate side liquid ports and use a 90° elbow (included) on the top of the drum. Self-priming pumps handle liquids with particulate matter at pressures up to 100 psi. Dead head operation allows discharge line to be closed with no damage or wear to the pump. Drum inlet connection is a 2" bung.
Minimal parts contact is excellent for use with shear-sensitive fluid. The PTFE diaphragms maintain their integrity when pumping highly aggressive fluids. Thermoplastic olefin (TPO) diaphragms maintain flexibility when used with acids or caustic fluids.
What's Included:
90° elbow; bung adapter; polypropylene (PP) pumps include a 33"L PVC suction pipe; Kynar
®
(PVDF) pumps include a 33"L PVDF suction pipe.
Specifications
Product Type
Air-Operated Drum Pumps
Air inlet pressure (psig)
100
Wetted parts
Polypropylene; PTFE diaphragm, check balls and O-rings
Max temperature (º C)
180
Motor (type)
Air-operated
Port size
1/2" NPT(F)
Brand
Kynar
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Positive Displacement Pump Types
Closed System Technology Drives the Trend Toward Safer, More Cost-Efficient Chemical Dispensing
How to Read a Pump Curve
Show All...
Guide to Liquid Pumps
Liquid Pump Installation Information
Liquid Pump Selection Flowchart
Liquid PumpsViscosity Handling Characteristics
Motor Enclosure Terminology
InnoCal Pump Reconditioning Program
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title