Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Alconox cleaner for manual or ultrasonic cleaning, 4 lb box

SI-17775-05
$35.00 USD/EACH

  • Order new bulk packages for added convenience
White powder Alconox®, Alcojet®, and Terg-A-Zyme® cleaners rinses off completely—with no residue. Alconox® cleaner is biodegradable; Alcojet® cleaner is low-sudsing; Terg-A-Zyme® cleaner combines the original Alconox® power with protein enzymes. Liqui-Nox® cleaner is biodegradable—use to remove soil, grease, blood, chemicals, and solvents. All cleaner are safe, odorless, and FDA certified.
ALCOJET, ALCONOX, LIQUI-NOX, TERG-A-ZYME-Reg TM Alconox Inc.
Product Type Equipment Cleaner
Format Powder
Packaging 4 lb box
Uses Manual or ultrasonic cleaning
Brand Alconox
Manufacturer number 1104-1
Model 1104-1





Technical Resources
  • Alconox MSDS.pdf (54 KB)
  • Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    Customers Who Bought This Item Also Bought
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title