dummy
EW-10508-74
- Measures velocity and temperature
- Sweep mode for one overall measurement
- Log, store and recall data
- Download data to a PC
-
- Optional telescopic handle available
The Model RVA501 is a data logging hand held
digital rotating vane anemometer used for air velocity
and volumetric flow measurements.
Also includes USB cable and logging software.
What's Included:
Hard carrying case, batteries, NIST-traceable calibration report supplied by manufacturer, and user manual.
| Product Type |
Vane Anemometer |
| Brand |
Alnor |
| Model |
RVA501 |
| Manufacturer number |
RVA501 |
at $7.50 USD/PACKAGE of 4
In stock.
at $155.00 USD/EACH
Usually ships in 8 days.
at $235.00 USD/EACH
Call 1-888-358-4705 for availability.
at $225.00 USD/EACH
Usually ships in 13 days.
Technical Resources
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection