United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 2
of
7 >
EW-01017-51
Representative Photo Only. Aluminum Crimpled-Walled Dishes with Tab, 43mm Dia x 1/2
Click to enlarge
More Product Details
Specifications
Related products
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
Recommended Products
dummy
Close[X]
EW-01017-51
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Balances & Scales
>
Balances / Scales Accessories
>
Weighing Dishes
>
Aluminum Weighing Dishes - Crimped Wall with Tab
Share
|
Aluminum Crimpled-Walled Dishes with Tab, 43mm Dia x 1/2" D, 1 gr., 20 ml Capacity, 1000/cs
EW-01017-51
$93.00 USD/CASE of 1000
In stock.
Qty
:
Add to Cart
Save to shopping list
Crimpled walled with side finger tab handle
Disposable
Crimpled-walled dishes with tab
Crimped sides for rigidity, sturdy gauge for heavy duty use. Large tab for sample numbers, flat bottom to improve sample handling, sides are angled for easy seperation. No oil residue, no vinyl coating to create contamination problems. Smooth bottom and may be used in a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical, and pharmaceutical.
Specifications
Product Type
Weighing Dish
Capacity (mL)
20
Material
aluminum
Quantity
1000/cs
Dimensions
2" dia x 1/2" D (5.1 x 1.3 cm)
Manufacturer number
D43-100
Model
43 mm
Related products
Qty
/
Price
Availability
Cole-Parmer SpatulaBalance
, 300 g x 0.1 g
EW-01019-11
at $43.00 USD/EACH
In stock.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Weighing Unit Conversion Table
Common Balance Terms
Equipping a Laboratory: A Study in Customer Service Excellence
Show All...
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Customers Who Bought This Item Also Bought
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title