Applikon Low-Well MicroFlask Systems Microtiter 96-well low-well polystyrene plate, pre-sterile, round bottom, 0.1 to 0.2 mL; bacterial mutant and construct libraries application
EW-29208-15
$7.00 USD/EACH
Are you a reseller or buying in bulk? Call us today.
Usually ships in 5 days.
Qty
  • Increase your incubator and shaker productivity without sacrificing culture quality!
  • System will accept any universal size 24- or 96-well plate.
  • Significantly reduce your evaporation loss rates in your samples to less than 2%
  • Improve consistency for all wells on titer plates when compared to standard titer plate experiments
  • Versatile system accepts 24/96 low-well plates or deep-well microtiter plates
  • Improve gas transfer in microtiter cultures
Replace roller bottles and shake flasks with these systems! The MicroFlask is used for cell culture and mircobial cultivation and uses microtiter plates for easy operation, flexibility, and reproducible results. The benefits of this system include no well-to-well cross-contamination, low and uniform evaporation rates, well-to-well reproducibility and excellent gas transfer to each culture. Stackable design allows for up to four plates under each clamp, allowing flexibility for your research.
For microbial cultures, bioreactors with microbial cultivations can require an air supply of up to two working volumes per minute. The MicroFlask sandwich covers allow for a headspace refreshment rate of 1 to 2 culture volumes per minute. This refreshment rate secures oxygen concentrations of at least 18% (v/v), even with oxygen uptake rates of 40 mmol 02/L-1 min-1.
For cell cultures, the MicroFlask permits optimal gas transfer, and also minimizes evaporation from your plate. You can grow cells several weeks in a single plate without contamination! Deep-well plates constructed of high-purity polystyrene are ideal for cell culture applications.
System setup is easy. The clamp mounts to your existing orgital shaking platform and securely holds microtiter plates in place. Orbital shaking platforms are ideal because this motion will increase the gas-liquid transfer, to provide uniformity in conditions across each well. Simply order a clamping device and sandwich covers (one cover per plate).
What's Included:
a gas-tight box for surrounding your plates with a custom atmosphere; excellent for biofuel applications! Use the cryo-replicator to sample from 96 frozen glycerol stocks. The glycerol stocks can remain frozen and viable for years. Use the replicator press for easy inoculation and replication of your cultures — ensuring accuracy during the replication sets of strains.
Product Type 96-well low-well, round bottom
Description Sterile, not reusable
Capacity (mL) 0.1 to 0.2
Use with Bacterial mutant and construct libraries
Model ZC3D003596
Brand Applikon
Accessories
Qty/Price
Availability
at $185.00 USD/EACH
Usually ships in 5 days.
at $310.00 USD/EACH
Usually ships in 5 days.
at $265.00 USD/EACH
Usually ships in 5 days.
at $212.00 USD/EACH
Call 1-888-358-4705 for availability.
at $212.00 USD/EACH
Call 1-888-358-4705 for availability.
at $212.00 USD/EACH
Call 1-888-358-4705 for availability.
Build a complete system
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  • 13-14 Spanish Catalog
  •  
    Service & Support
    Still have questions
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title