United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
4 >
Representative photo only
Applikon Microflask Standard Sandwich Cover for 24 well deep well plates and inlays approx 40 mmol L hr
Click to enlarge
More Product Details
Specifications
Accessories
Build a complete system
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-29208-09
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Life Science
>
Fermentation
>
Applikon Deep-Well MicroFlask Systems
>
Applikon Deep-Well MicroFlask Systems Sandwich Covers with Layers
Share
|
Applikon Microflask Standard Sandwich Cover for 24-well deep-well plates and inlays, approx. 40 mmol/L/hr
EW-29208-09
$180.00 USD/EACH
Usually ships in 5 days.
Qty
:
Add to Cart
Save to shopping list
Increase your incubator and shaker productivity without sacrificing culture quality!
Significantly reduce your evaporation loss rates in your samples to less than 2%
Improve consistency for all wells on titer plates when compared to standard titer plate experiments
Versatile system accepts 24/96 low-well plates or deep-well microtiter plates
Improve gas transfer in microtiter cultures
Replace roller bottles and shake flasks with these systems! The MicroFlask is used for cell culture and microbial cultivation and uses microtiter plates for easy operation, flexibility, and reproducible results. The benefits of this system include no well-to-well cross-contamination, low and uniform evaporation rates, well-to-well reproducibility and excellent gas transfer to each culture. Stackable design allows for up to four plates under each clamp, allowing flexibility for your research.
For microbial cultures,
bioreactors with microbial cultivations can require an air supply of up to two working volumes per minute. The MicroFlask sandwich covers allow for a headspace refreshment rate of 1 to 2 culture volumes per minute. This refreshment rate secures oxygen concentrations of at least 18% (v/v), even with oxygen uptake rates of 40 mmol 0
2
/L
-1
min
-1
.
For cell cultures,
the MicroFlask permits optimal gas transfer, and also minimizes evaporation from your plate. You can grow cells several weeks in a single plate without contamination! Deep-well plates constructed of high-purity polystyrene are ideal for cell culture applications.
System setup is easy. The clamp mounts to your existing orgital shaking platform and securely holds microtiter plates in place. Orbital shaking platforms are ideal because this motion will increase the gas-liquid transfer, to provide uniformity in conditions across each well. Simply order a clamping device and sandwich covers (one cover per plate).
Sandwich Covers with Layers
These sandwich covers have re-usable layers. They include a stainless steel lid for rigid support, a microfiber cotton layer, ePTFE (0.3 µm pores) layer for sterility, and a soft silicone layer to seal the system. All components can handle repeated autoclaving.
What's Included:
a gas-tight box for surrounding your plates with a custom atmosphere; excellent for biofuel applications! Use the cryo-replicator to sample from 96 frozen glycerol stocks. The glycerol stocks can remain frozen and viable for years. Use the replicator press for easy inoculation and replication of your cultures ensuring accuracy during the replication sets of strains.
Specifications
Product Type
Sandwich Cover
Cover type
Standard
Well size
24-well deep-well
Oxygen transfer rate (mmol/L/hr)
about 40
Orbital shaking frequency (rpm)
300
Shaking amplitude (mm)
≥ 50
Brand
Applikon
Build a complete system
1
.
Applikon Deep-Well MicroFlask Systems Clamping Devices
Details
2
.
Applikon Deep-Well MicroFlask Systems Microtiter Plates
Details
3
.
Applikon Deep-Well MicroFlask Systems Sandwich Covers with Layers
Details
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Cole-Parmer Introduces Enhancements to Life Science Offering
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title