Lovibond L147490 Test Kit, Chlorine (Sodium Hypochlorite), 2 to 18% NaOCl
Representative image only.
Lovibond Mfr # 147490 – Item # EW-05542-36
Accurate and easy to use for routine water analysis
  • Continuous color disc
  • Tablet and powder reagent chemistries
  • 1 year warranty
PRODUCT OPTIONS
$135.00USD / Each
$150.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • TestSodium hypochlorite, ultra-high range
  • Measurement Points2 to 18% NaOCl
  • Tests Per Kit100
  • DescriptionTest Kit , Chlorine (SOD ium Hypochlorite), 2-18% Naocl
  • Warranty1 year
1 Year Warranty
Key Features
  • Continuous color disc
  • Tablet and powder reagent chemistries
  • 1 year warranty
More About this Item
Simply add the tablet or powder pack reagent to the sample cell, allow it to dissolve per specific parameter instructions, rotate the color disc to match the color of the sample, and read off the concentration value.