dummy
EW-94101-00
Call 1-888-358-4705 for availability.
Qty:
- Use in the laboratory or out in the field!
- Greaseless glassware provides quick and effortless assembly and disassembly
- Built-in high-speed printer provides hard copy results and duplicates if required
The Cou-Lo Aquamax titrator sets new standards in versatility and ease of operation for determining water content in liquids, gases, and powders. The patent-pending ACE (Automatic Compensation of Errors) control system guarantees that the actual electrolysis current produced and the count rate displayed are always correctly synchronized, regardless of changes to the electrolysis cell resistance. This system provides the correct result even if the generator is partially blocked, minimizes errors in daily calibration, and increases the number of samples you can analyze. Use for ASTM D1533, D4928, D6304, IP 386, API MPMS Chapter 10.9, and BS 60814 test methods.
A truly versatile instrument, the Cou-Lo Aquamax displays results in %, ppm, mg/kg, and µg of water. Delayed start, automatic three-fold end point indication, and four different calculation modes accommodate a variety of applications. Printed results show statistics, data input parameters, sample ID numbers, and time-and-date stamp. Results can also be downloaded via the optional Results Manager software package into a PC spreadsheet format.
What's Included:
built-in stirrer, titration vessel, detector electrode, generator electrode, drying tube, injection septa, glass syringe, luer needle, molecular sieve, stir bar, funnel, printer, paper, universal AC voltage adapter, DC car adapter, and internal battery.
| Product Type |
Karl Fischer Titrators |
| Range |
1µg to 100 mg water |
| Resolution |
0.1µg |
| Display |
LCD |
| Display resolution |
40-characters |
| Units |
%, ppm, mg/kg, µg |
| Accuracy |
From 10 to 100µg: ±3 µg; From 100 µg to 1 mg ±5 µg; Above 1 mg: ±0.5% |
| Dimensions |
9.875 in L x 9.75 in W x 4.75 in H |
| Power (VAC) |
90 to 264; 12 VDC car adapter |
| CE Compliance |
Yes |
| Power (Hz) |
47 to 63 |
| Brand |
Aquamax |
| Manufacturer number |
71000 |
| Model |
71000 |
at $1,200.00 USD/EACH
Call 1-888-358-4705 for availability.
at $1,199.30 USD/EACH
Call 1-888-358-4705 for availability.
at $2,798.35 USD/EACH
Call 1-888-358-4705 for availability.
at $136.75 USD/EACH
Usually ships in 13 days.
at $90.00 USD/EACH
Usually ships in 13 days.
at $84.20 USD/EACH
Usually ships in 13 days.
at $273.55 USD/PACKAGE
Call 1-888-358-4705 for availability.
at $1,472.80 USD/EACH
Call 1-888-358-4705 for availability.
at $526.00 USD/EACH
Call 1-888-358-4705 for availability.
Technical Resources
Virtual Catalog (PDFs)
More info
Related technical articles (Roll over for summary)
Show Less...
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.