log in or register
Aquamax Volumetric Karl Fischer Titrator
EW-94100-01
$6,150.25 USD/EACH
Usually ships in 7 days.
Qty
  • Versatile instrument for the volumetric determination of water content using the KF method
  • Very easy to use - function guide display walks you through operation
  • Novel syringe control eliminates the common problem of bubble formation in the reagent circuit
User-friendly interface and intuitive keypad make operation of the Aquamax Volumetric KF Titrator simple to grasp. Titrator features two high-accuracy 5-mL syringes with 40,000 step resolution for superior dosage accuracy. One syringe can be used for the KF reagent and the other can be used for dosing of standard titre, different factor reagents, or specialty reagents. Flexible titrator accepts several different reagent brands.
Titrator is programmable and stores up to 10 different titration and standardization programs. Automatically monitor and compensate for drift during titration - pop up annunciator will prompt the user if drift exceeds programmed limit values. Built-in datalogging capability stores up to 55 analysis results. Optional communications software or printer allow for the viewing or printing of stored data for data management flexibility.
What's Included:
two glass syringes, two burette inlet tubes, two burette outlet tubes, five drier cartridges, inlet pump tube with bottle cap, two inlet and outlet tubes with conical adapters, outlet pump tube, magnetic stirrer and titration vessel, electrode, electrode cable, ten O-rings, and universal power adapter.
Product Type Volumetric Titrators
CE Compliance Yes
Brand Aquamax
Manufacturer number 91000
Accessories
Qty/Price
Availability
at $421.00 USD/EACH
Call 1-888-358-4705 for availability.
at $526.00 USD/EACH
Call 1-888-358-4705 for availability.
Thermal Printer
EW-94100-27
at $1,019.75 USD/EACH
Call 1-888-358-4705 for availability.
TiCom Software kit
EW-94100-33
at $1,877.50 USD/EACH
Call 1-888-358-4705 for availability.
Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Leasing Available
    Financing is avaliable for many of our products.
    See Details
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title