Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Arc Flash Protective Flash Suit Kits, 40 cal/cm2, double extra large

EW-86436-83
$1,167.00 USD/EACH

Usually ships in 10 days.
Qty

  • Protective suits meet NFPA 70E hazard risk category (HRC) 4
Constructed of Indura Ultra Soft® (88% cotton) for comfort and breathability, these arc flash suits are rated for 40 cal/cm2 Arc Thermal Protective Value (ATPV). Jackets feature 32" (81.3 cm) length, Nomex® wristlets, and hook and pile storm closure. Bib overalls feature 20" (76.2 cm) inseam, integrated heavy duty suspenders, and hook and pile waist straps. Pro-Hood™ is made of Indura Ultra Soft with a 10" x 20" (25.4 x 50.8 cm) anti-fogging lens. Hood meets ASTM F2178 and NFPA 70E standards along with the flammability requirements of ASTM F1506; also CE 0120 certified.

Compliance: Coat and bib overalls meet ASTM 1506 and NFPA 70E standards and are CE 0120 certified.
What's Included:
arc flash coat, bib overalls, Pro-Hood, hard hat, safety glasses, and a carrying bag.

Product Type Arc Flash Protection
Description Suit Kit
Size XX-Large
Color Gray
Material Indura Ultra Soft - Standard weight
Compliance Hazard Risk Category 4
Length (in.) 32
Quantity 1/ea
Manufacturer number SK40-2XL
Brand Salisbury
Model SK40-2





Technical Resources
Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title