- Protective suits meet NFPA 70E hazard risk category (HRC) 4
Constructed of Indura Ultra Soft® (88% cotton) for comfort and breathability, these arc flash suits are rated for 40 cal/cm2 Arc Thermal Protective Value (ATPV). Jackets feature 32" (81.3 cm) length, Nomex® wristlets, and hook and pile storm closure. Bib overalls feature 20" (76.2 cm) inseam, integrated heavy duty suspenders, and hook and pile waist straps. Pro-Hood™ is made of Indura Ultra Soft with a 10" x 20" (25.4 x 50.8 cm) anti-fogging lens. Hood meets ASTM F2178 and NFPA 70E standards along with the flammability requirements of ASTM F1506; also CE 0120 certified.
Compliance: Coat and bib overalls meet ASTM 1506 and NFPA 70E standards and are CE 0120 certified.
What's Included:
arc flash coat, bib overalls, Pro-Hood, hard hat, safety glasses, and a carrying bag.