United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
8 >
Representative photo only
Arc Flash Protective Jacket Overpant Kits 8 cal cm2 medium
Click to enlarge
More Product Details
Specifications
Technical Resources
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-86436-70
was succesfully added to your cart.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Safety Products
>
Arc Flash Products
>
Arc Flash Apparel
>
Salisbury by Honeywell Arc Flash Protective Jacket and Overpant Kits
Share
|
Arc Flash Protective Jacket/Overpant Kits, 8 cal/cm2, medium
EW-86436-70
$365.00 USD/EACH
Usually ships in 10 days.
Qty
:
Add to Cart
Save to shopping list
Kits include jacket, overpants, helmet with face shield, safety glasses, and carrying bag.
Pro-Wear
Jackets and Overpants features
* Made with Indura Ultra Soft
®
(88% cotton) for comfort and breathability
* Meet current ASTM F1506 and NFPA 70E standards
* CE 0120 certified
* Jackets feature 32" length, Nomex
®
wristlets, and hook & pile front closure
* Overpants feature 30" inseam, hook & pile cuff adjustment, and loose design for fitting over existing work clothes
Helmet with face shield features
* Face shield has 70% visual light transmission (VLT) best in the industry!
* 7.5" x 20" viewing area
* Absorbs 99.9% of harmful UV radiation
* Tested to ASTM F2178 specifications
Specifications
Product Type
Arc Flash Protection
Description
Jacket & Overpant Kit
Size
Medium
Color
Black
Material
Indura Ultra Soft
Compliance
Hazard Risk Category 2
Length (in.)
32
Quantity
1/ea
Manufacturer number
SKJP8-M
Brand
Salisbury
Model
SKJP8-M
Technical Resources
Related technical articles (Roll over for summary)
Technical Expertise: How Cole-Parmer Application Specialists Respond to Customers' Needs
ABCs of DMMs
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title