United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
1
of
5 >
EW-68840-00
Ashcroft
®
Deadweight Tester Kit, 1000 psi Maximum
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-68840-00
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17101-09
NIST-Traceable Calibration; Hydraulic Deadweight Tester, 0.1% single column
$400.00 USD/EACH/EACH
Usually ships in 8 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Pressure & Vacuum Instruments
>
Calibrators
>
Ashcroft Deadweight Tester kits
Share
|
Ashcroft Deadweight Tester Kit, 1000 psi Maximum
EW-68840-00
$4,462.00 USD/EACH
Call 1-888-358-4705 for availability.
Qty
:
Add to Cart
Save to shopping list
Primary pressure standard accurate to ±0.1% of reading
These self-contained tester kits have everything you need for testing, setting, calibrating, and repairing your high-accuracy pressure instruments—except operating fluid. Testers come with a certificate verifying traceability to NIST-traceable standards.
Each kit consists of a two-stage hydraulic pump, two interchangeable piston assemblies (high- and low-pressure), a manifold with connectors, and an assortment of nonmagnetic, die cast zinc alloy weights. Precision-tooled weights are premeasured, marked with pressure values for each piston and come with a separate case.
Use the high-pressure piston for calibration pressures at maximum psi with a 75 psi resolution (see table at below); use the low-pressure piston to achieve 1/5 of the maximum pressure with a 15 psi resolution.
During operation, the manifold is pressurized until the weight assembly reaches a designated mark. A displacement valve lets you vary the pressure without using the pump handle or release valve. Once the correct pressure is achieved, simply reposition the needle on your gauge to the corresponding value.
Max psi in the table is with a 75 psi resolution using the high-pressure piston.
Specifications
Product Type
Deadweight Testers
Description
Deadweight Tester
Piston/cylinder material
400-series stainless steel
Operating fluid
SAE 20 automotive or machine oil (approximately 1-1/2 pints required; not included)
Body material
aluminum alloy
Accuracy
±0.1% of reading
Pressure range
15 to 1000psi
Brand
Heise
Manufacturer number
1305D-10
Model
1305D
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Hydraulic Deadweight Tester, 0.1% single column
EW-17101-09
at $400.00 USD/EACH
Usually ships in 8 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Absolute, Gauge, and Differential Pressure Scales
Common Pressure Terms
Pressure Unit Conversion Constants
Show All...
The Hidden Costs of Mercury-Filled Vacuum Gauges
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title