United States
[Change]
Shopping Cart
Subtotal
=
$0.00
Items
=
0
View Cart
United States
1-800-323-4340
or
Live Chat
Welcome,
log in
or
register
Search
Shop All Products
Balances & Scales
Baths and Circulators
Calibration
Chemicals, Reagents, & Lubricants
Clean Room
Cooling Equipment
Fittings
Flow, Level & Valves
Fluid & Liquid Handling
Furniture & Fume Hoods
Heating Equipment
Industrial Equipment
Lab Equipment
Laboratory Cleaning Supplies
Laboratory Supplies & Consumables
Life Science
Masterflex Peristaltic Pumps
Masterflex Pump Tubing
Material Handling and Storage
Material Testing & Physical Testing
Mixers and Blenders
pH Meters and Water Quality Products
Pressure & Vacuum Instruments
Process Equipment & Instrumentation
Pumps
Recorders & Data Acquisition
Safety Products
Shop by Industry
Spectroscopy and Spectrophotometers
Temperature, Humidity, Moisture and Weather Products
Tubing, and Hose
Shop by
|
Service & Support
|
Technical Resources
|
My Account
Check order status
View Product List
< 3
of
4 >
EW-02940-04
High-Precision Abbe refractometer; 0 to 95.0% Brix, 1.3000 to 1.7000 RI
Click to enlarge
More Product Details
Specifications
Calibrations
Technical Resources
Virtual Catalog (PDFs)
Related technical articles
Service & Support
Answer Box -Q&A posts
Product Reviews
dummy
Close[X]
EW-02940-04
was succesfully added to your cart.
Would you like it calibrated?
Price
:
Availability
:
EW-17105-00
NIST-Traceable Calibration; Refractometer
$75.00 USD/EACH/EACH
Usually ships in 9 days.
No Calibration Needed
Yes, Add Calibration
Learn more:
Why calibrate Test Equipment?
Home
>
Lab Equipment
>
Refractometers
>
Abbe Refractometers
>
Atago Abbe Refractometers
Share
|
Atago High-Precision Abbe refractometer; 0 to 95.0% Brix, 1.3000 to 1.7000 RI
EW-02940-04
$7,700.00 USD/EACH
In stock.
Qty
:
Add to Cart
Save to shopping list
Uncompromising precision at an economical cost
Meets ASTM D 1416-89, Section 38-46 requirements
These Abbe refractometers provide highly accurate refractive index and Brix measurements of liquids and solids. Place your sample on the main prism, which is rimmed with stainless steel to prevent corrosion, look through the eyepiece and line up the shadowline in the crosshairs. The built-in 589 nm illuminator light source provides stable, monochromatic light (02939-00, -02 include LED light source), making it easy to read the black lines on the green field. The frosted secondary prism directs light to the main prism evenly, and lifts up for easy placement of solids and viscous liquids on the main prism. The color compensator knob allows you to adjust the boundary line for higher clarity and fine tuning capability for greater resolution. Use the color compensation knob and a conversion table to measure dispersion (variance of RI with different wavelengths of light).
In order to maintain constant temperature at both prisms, these refractometers include four 9-mm OD ports (using 6 to 7-mm ID tubing) for connections to a constant-temperature bath. A thermistor thermometer for measuring temperature is built into the main prism. The temperature of the main prism is indicated on the LCD from 0 to 50°C with ±0.2°C accuracy; a digital temperature display unit is included. The built-in desiccant chamber removes moisture from internal optics ensuring an unobstructed view of scales.
Model 02939-04
is a high-precision Abbe that measures both Brix and refractive index of liquids, solids, and semisolids. Unit features an accuracy pf ±0.05% Brix and ±0.0001 RI - perfect for meeting your precision quality requirements.
What's Included:
a digital temperature display unit, standard solution (4-mL of monobromonaphthalene), a screwdriver for scale adjustment, and a 5-ft cord with three-prong plug.
Specifications
Product Type
Abbe Refractometer
Brix scale range (Minimum)
0
Brix scale range (Maximum)
95
Refractive Index (Minimum)
1.3
Resolution
0.1% Brix. 0.0002 RI
Refractive Index (Maximum)
1.7
Temp accuracy
±0.2°C
Accuracy
±0.05%, ±0.0001
Sample volume
0.2 mL
Dimensions
4.75 in W x 13.375 in H x 12.25 in D
Model
1230 3T
Manufacturer number
1230 3T
Display
3-digit LCD
Power
115 VAC, 50/60 Hz
Calibrations
Qty
/
Price
Availability
NIST-Traceable Calibration; Refractometer
EW-17105-00
at $75.00 USD/EACH
Usually ships in 9 days.
Add to Cart
Technical Resources
Virtual Catalog (PDFs)
13-14 Catalog
Related technical articles (Roll over for summary)
Testing Refractive Index of Candy
Refractometers
Brix Scale (sucrose)
Show All...
Equipping a Laboratory: A Study in Customer Service Excellence
Electron zap turns flimsy plastic into sturdy shrink wrap
Show Less...
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Leasing Available
Financing is avaliable for many of our products.
See Details
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here.
Call
1-800-323-4340
Option
9
ext.
6334
See Details
Questions and Answers
Product Reviews
Close
Save My Shopping List
Please login in order to use Shopping Lists
Close
Close
Save My Shopping List
You can
view your shopping list
or
continue shopping
View products by:
Title
|
Brand
|
Category
|
Price
|
Popularity
|
Best Sellers
View product categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Title
|
Product Category
|
Recommended
View popular items by:
Popularity
|
Favorites
|
Title
|
Price
|
Part Number
|
Category
View Popular Categories by:
Popularity
|
Our Choices
|
All-Around Favorites
|
Product Category
|
Recommended
|
Title