Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Bacdown Hand Soap, 5 L

EW-86115-10
$89.00 USD/EACH

    Gentle handsoap combines soothing, coconut-based surfactants with skin-kind Triclosan to provide a handsoap that may be used repeatedly with minimal skin drying. It is recommended for applications where repeated hand scrubbing is necessary.
    Product Type Hand Cleaner
    Format Pump
    Volume (Liters) 5
    Packaging 5 L container
    Brand Bacdown
    Manufacturer number 7005
    Model 7005





    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title