| Frequency range |
±5% = 180 to 480,000 RPM (3 to 7,000 Hz); ±3 dB = 24 to 840,000 RPM (0.4 to 14,000 Hz) |
| Vibration range (Velocity (peak)) |
0.001 to 199.9 in./sec (peak) |
| Vibration range (Acceleration) |
0.001 to 199.9 gs (peak) |
| Display |
Graphic LCD with low-battery indicator |
| Vibration range (Displacement (peak to peak)) |
0.015 to 199.9 mils (peak-to-peak) |
| Dimensions |
7.67" H x 3.97" W x 1.73" D (19 x 10 x 4 cm) |
| Power |
Four AA batteries (included) |
| Accuracy |
5% of full scale |
| Brand |
Balmac |
| Model |
230M |