Item Total = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Balston Vacuum Pump Exhaust Filter, 9 cfm, 3/4 NPT(F); 1/Pk

Item# EW-29180-20

$214.00 USD/EACH

Are you a reseller or buying in bulk? Call us today.


Available for order
or call to order
Qty

  • Eliminate 99% of oil mist and smoke from vacuum pump exhaust
  • Prevent oil accumulation in ductwork
  • Eliminate potential OSHA and EPA violations
  • Protect vacuum pumps and reduce costly maintenance
Balston Vacuum Pump Exhaust Filters remove all visible oil mist and smoke from vacuum pump exhaust, even if/when saturated with oil. The high-efficiency filter cartridge continuously drains the collected fluid, allowing for expensive lubricating fluid to be collected.
This exhaust filter's rating should be equal to or greater than that of the free air capacity of the pump it is being connected to.
Housing material Steel
Product Type Inlet/outlet filter
Air flow rate (ft3/hr) 9 CFM
Connections 3/4" NPT
Max pressure 15 psig
Max temperature 180 oC
Applications For non-hazardous/non-corrosive applications
Qty/ea 1
Brand Balston





Technical Resources
 
Service & Support
Still have questions
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 8 ext. 138-8456
Questions and Answers
Product Reviews

REVIEW SNAPSHOT®

by PowerReviews
 
5.0

(based on 1 review)

Reviewed by 1 customer

Displaying review 1

Back to top

 
5.0

A Good Investment here

By Kenny

from LSU LA Emerging Tech

Verified Reviewer

Do not let the expense deter you from buying this filter. I am amazed by the elimination of exhaust smoke and oil drips around the vacuum pump on the floor. There is also no noise either while running our vacuum pump in our research lab which is used constantly for experiments. Simply said that eliminating the oil spray exhaust also means less need for adding new oil and cleanup of the floor and cabinets in our lab.

Displaying review 1

Back to top