Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Beijer iX QTERM-A12 Large 12.1" Graphics

EW-68516-10
$2,595.00 USD/EACH

Usually ships in 5 days.
Qty

  • State of the art touch screen technology driven by industry leading software
  • Wide operating temperature range of 22 to 158°F (30 to 70°C)
  • UL listed Class 1 Div 2 certified
  • IP66 and NEMA 4X sealing for harsh environments
  • Polymer housing
  • iX software for automation control or Windows® embedded CE 6.0 development
Higher user confidence in your application is gained with easy-to-read display and fast touch response. Easy mounting and access to connections save time for engineers.

The iX software at the core of these panels takes you quickly from ideation to a running system. Filled with smarter design engineering tools and solid HMI functionality, iX provides intuitive, friendly, but powerful HMI graphics for your applications in an open platform. It’s flexible and open for advanced programmers, yet easy to use and filled with ready-made features resulting in efficient, flexible project development for all. Unlimited iX Runtime software is included in this display. Also included are drivers to equipment from all major vendors to give you system flexibility.Choose from our offering of Human Machine Interface productsThese rugged QTERM® HMI panels are designed to perform in environments with high levels of shock, vibration, humidity and extreme temperature ranges. They are certified and listed accordingly for hazardous areas: IP66 and NEMA 4X sealing, UL 50 Type 4X, UL 508, and UL listed Class I Div 2 certified.

A wide range of drivers along with Ethernet 10/100Base-T, two USB 2.0 full-speed host ports, and two serial ports enables connection to nearly any device. The panel comes loaded with your choice of intuitive iX software for automation control or you can program directly in the Windows embedded CE 6.0 environment.
What's Included:
Hardware packet for mounting unit into panel.
Display 12.1 in. (307 mm) diagonal
Interface Ethernet 10/100Base-T, RJ-45, One EIA-232/422/485 DB9f; one EIA-232 DB9f, Two USB 2.0 full-speed host port (type A connectors)
Panel cutout (" W) 9.8
Panel cutout (" H) 7.4
Power (VDC) 10 to 32
Touch screen Analog-resistive with 4H hardness
Brand BEIJER
Model QTERM®-A12
Display dimensions 13.5 in. x 10.5 in. x 2.7 in. (344 x 267 x 69.1 mm)
Memory 128 MB RAM, 4 GB flash
Display type 800 x 600 SVGA, TFT color LCD
Processor XScale™ PXA 300 ARM 624 MHz
Front panel rating IP66, NEMA 4X
Rear panel rating IP66, NEMA 4X
Manufacturer number IX QTERM-A12





 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title