SHOWA Best Hustler Chemical-Resistant Gloves; size, large, 12" gauntlet; 12 pair
Representative image only.
SHOWA Mfr # 725R-10 – Item # EW-86288-12
SPECS & DESCRIPTION
Specifications & Description
  • SizeLarge (9-10)
  • Length (in)12
  • Length (cm)30.5
  • ColorRed
  • SterileNo
  • DescriptionBest Hustler Chemical-Resistant Gloves; size, large, 12\" gauntlet; 12 pair
More About this Item
PVC-coated and nitrile reinforced gloves are abrasion-resistant and provide protection from solvents and acids. Two-piece interlock liner enhances flexibility and comfort. Rough, sanded-on finish ensures a secure grip when working with oil or grease—ideal for petrochemicals refining and material handling. They are SANITIZED® and launderable. Pack of 12 pair.