log in or register
Beta cell; includes removable cell mount, 12 assorted PTFE spacers, slide plate holder, and instructions
EW-83001-00
$73.00 USD/EACH
Usually ships in 7 days.
Qty
    Ideal for smears, this economical cell is easy to assemble. Simply place sample between two plain windowa separated by spacers, place in holder, and install threaded retainer until assembly is sealed. Requires two NaCl windows (sold separately).
    Accessories
    Qty/Price
    Availability
    at $33.00 USD/EACH
    In stock.
    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title