log in or register
Beta press, Includes barrel, bolts, slide holder, and instructions, for use with Scanning Infrared Spectrophotometer
EW-83002-00
$105.50 USD/EACH
Usually ships in 12 days.
Qty
    This unique precision solid sampling device makes KBr pellets in a 316 stainless stell barrel with two hardened and highly polished 316 stainless steel bolts. Remove bolts and mount beta press directly onto the spectrophotometer for analysis.
    Accessories
    Qty/Price
    Availability
    at $45.00 USD/EACH
    Call 1-888-358-4705 for availability.
    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title