Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Biohazard Bag 14" x 19", 200 per Case

EW-06494-00
$119.00 USD/CASE

    Bright orange-red, 1.25 mil, high-molecular weight HDPE bags are printed with the biohazard warning symbol and precautionary procedures in four languages. The sterilization indicator patch darkens when autoclaved. Autoclavable at 250°F (121°C).
    Order stands and plastic ties separately.
    Product Type Biohazard Bags
    Material HDPE
    Capacity (gal.) 4
    Size (" W) 14
    Size (" L) 19
    Thickness (mil) 1
    Color Orange and Red
    Manufacturer number F13164-1419

    Accessories
    Qty/Price
    Availability
    Plastic tie, 4"L
    EW-06831-15

    at $15.75 USD/PACKAGE of 1000

    In stock.




    Technical Resources
    Virtual Catalog (PDFs)
  • 13-14 Catalog
  •  
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title