Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register
 

Biohazard Waste Can, 10 gallon

EW-81772-22
$84.00 USD/EACH

Usually ships in 4 days.
Qty

  • Highly visible — alerts workers to potential biohazards
Made of heavy-duty polyethylene. Open lid by hand or with foot pedal. Includes prominent biohazard warning label.
Approval: FM
Product Type Safety Cans
Capacity (Liters) 38
Capacity (gal.) 10
Dimensions (in.) 22 in L x 18 in W x 18 in H
Brand Eagle
Model 945BIO





 
Service & Support
Still have questions?
Contact Our Experts
for FREE product
application selection
and support.
or call 1-800-323-4340
Custom Ordering Solutions
Custom Ordering Solutions
Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
Questions and Answers
Product Reviews
View products by: Title | Brand | Category | Price | Popularity | Best Sellers
View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title