Subtotal = $0.00 
Items = 0 
View Cart
United States
1-800-323-4340 or 
Welcome,
log in or register

Bismuth Powder -100 Mes 100g

EW-88289-97
$56.00 USD/EACH

    A lab reagent is a staple in chemical applications. It is a great way to test chemical reactions or to see if a reaction will occur. You can find one just about anywhere including Cole-Parmer’s catalog which features a broad range of laboratory reagents, cleaning chemicals and buffers commonly found in labs. But occasionally workers run across an application that requires a specialty lab reagent. You might need Malachite Green, a high purity biological stain. How do you go about selecting a lab reagent especially if there are new and more advanced compounds out there that you might want to try? For the novice a lab reagent kit may be in order. Using kits can prevent mistakes that are related to the reagent rather than the application. If you’re pretty advanced and haven’t found that specialty compound you’re searching for try talking to a Cole-Parmer technical expert. They often have information about chemicals not listed on the website but available for special order or by other means. Cole-Parmer also offers custom blending services for specific chemical mixes. With advances in technology selecting a lab reagent may not be as routine as originally thought so it’s great to have some help.

    This item is considered a hazardous material.Please note: there may be additional shipping restrictions like freight collect or no priority air service, extra freight charges may apply. This item(s) may be restricted from shipping to a residential address, sold to private individuals or exported from the United States. For additional information or questions please call us at 1-800-323-4340 (US) or 847-549-7600 (International).
    Product Type Chemical Reagents
    Form Powder
    Manufacturer number AC318091000





    Technical Resources
     
    Service & Support
    Still have questions?
    Contact Our Experts
    for FREE product
    application selection
    and support.
    or call 1-800-323-4340
    Custom Ordering Solutions
    Custom Ordering Solutions
    Let us find the exact product you need. We have access to suppliers full lines - for products beyond what you see here. Call 1-800-323-4340 Option 9 ext. 6334
    Questions and Answers
    Product Reviews
    View products by: Title | Brand | Category | Price | Popularity | Best Sellers
    View product categories by: Popularity | Our Choices | All-Around Favorites | Title | Product Category | Recommended
    View popular items by: Popularity | Favorites | Title | Price | Part Number | Category
    View Popular Categories by: Popularity | Our Choices | All-Around Favorites | Product Category | Recommended | Title